Product Details

SNP ID
rs78619382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26501023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGTGTTATGCCTCACAGTTCC[C/T]TTCTCCTAAGTTTTAGAGTTAGAGT
Phenotype
MIM: 601610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTN1A1 PubMed Links

Gene Details

Gene
BTN1A1
Gene Name
butyrophilin subfamily 1 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001732.2 Intron NP_001723.2
XM_005249340.3 Intron XP_005249397.2

View Full Product Details