Product Details

SNP ID
rs12276189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124753139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGCAACTTGTTTCTTATCTCAT[A/G]CAAACATTATGTATCTTTATTTAAA
Phenotype
MIM: 614281 MIM: 602350 MIM: 606011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESAM PubMed Links

Gene Details

Gene
ESAM
Gene Name
endothelial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138961.2 1274 UTR 3 NP_620411.2
XM_005271720.4 1274 UTR 3 XP_005271777.1
XM_011543060.2 1274 UTR 3 XP_011541362.1
Gene
LOC101929340
Gene Name
uncharacterized LOC101929340
There are no transcripts associated with this gene.

Gene
NRGN
Gene Name
neurogranin
There are no transcripts associated with this gene.

Gene
VSIG2
Gene Name
V-set and immunoglobulin domain containing 2
There are no transcripts associated with this gene.

View Full Product Details