Product Details

SNP ID
rs73491506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48789520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATCCCACAGGTCTCAGCTCAGC[T/C]CCAGCCCATCACCACCCCCAATACA
Phenotype
MIM: 305371
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GATA1 PubMed Links

Gene Details

Gene
GATA1
Gene Name
GATA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002049.3 Intron NP_002040.1
XM_011543897.2 Intron XP_011542199.1
XM_011543898.2 Intron XP_011542200.1

View Full Product Details