Product Details

SNP ID
rs16981364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:88753098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAAGGAAATAATTGGGCCTTCT[A/C]TTCAGAACTAAATGTGTTCTAGTGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPXCR1 PubMed Links

Gene Details

Gene
CPXCR1
Gene Name
CPX chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184771.1 Intron NP_001171700.1
NM_033048.5 Intron NP_149037.4

View Full Product Details