Product Details

SNP ID
rs77740406
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:111474110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGTCGTCTAATTGTTAGTGG[A/T]TTCTATCACTTGTTTTCTTTGATGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf162 PubMed Links

Gene Details

Gene
C1orf162
Gene Name
chromosome 1 open reading frame 162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300834.1 Intron NP_001287763.1
NM_001300835.1 Intron NP_001287764.1
NM_174896.3 Intron NP_777556.1
XM_011540712.2 Intron XP_011539014.1
XM_017000323.1 Intron XP_016855812.1
XM_017000324.1 Intron XP_016855813.1
Gene
TMIGD3
Gene Name
transmembrane and immunoglobulin domain containing 3
There are no transcripts associated with this gene.

View Full Product Details