Product Details

SNP ID
rs9442945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73696029 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGCGCCATTGTAATGGGGATG[A/G]GAGGGGTGGAGCCTCCAAGTCCTGT
Phenotype
MIM: 608859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD109 PubMed Links

Gene Details

Gene
CD109
Gene Name
CD109 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159587.2 Intron NP_001153059.1
NM_001159588.2 Intron NP_001153060.1
NM_133493.4 Intron NP_598000.2
XM_005248659.3 Intron XP_005248716.1
XM_011535473.1 Intron XP_011533775.1
XM_017010283.1 Intron XP_016865772.1
Gene
LOC101928489
Gene Name
uncharacterized LOC101928489
There are no transcripts associated with this gene.

View Full Product Details