Product Details
- SNP ID
-
hCV27848205
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12957022 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGACCACCCCGGAGGCGGCGCCG[A/T]AACGCGCGAACTGCTTAGCCGCGTA
- Phenotype
-
MIM: 605162
MIM: 600061
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GADD45GIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140839998] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GADD45GIP1
- Gene Name
- GADD45G interacting protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052850.3 |
233 |
Missense Mutation |
TAC,TTC |
Y64F |
NP_443082.2 |
- Gene
- RAD23A
- Gene Name
- RAD23 homolog A, nucleotide excision repair protein
There are no transcripts associated with this gene.
View Full Product Details