Product Details
- SNP ID
-
rs73193201
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126986049 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCTGGAGTTTGGATGGTGGCT[C/T]CTTAGCCTGGTCCCTCCTGTTGAAT
- Phenotype
-
MIM: 601055
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLXNA1
PubMed Links
Gene Details
- Gene
- PLXNA1
- Gene Name
- plexin A1
View Full Product Details