Product Details

SNP ID
rs183338110
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:33081377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCTGCCTCCCTGCCCCTTCTG[C/T]CTCCTGTCAGCAGAGGCCTCGGCTC
Phenotype
MIM: 608353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AZIN2 PubMed Links

Gene Details

Gene
AZIN2
Gene Name
antizyme inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293562.1 249 Intron NP_001280491.1
NM_001301823.1 249 UTR 5 NP_001288752.1
NM_001301824.1 249 UTR 5 NP_001288753.1
NM_001301825.1 249 Intron NP_001288754.1
NM_001301826.1 249 Intron NP_001288755.1
NM_052998.3 249 Intron NP_443724.1
XM_005270404.2 249 Intron XP_005270461.1
XM_005270406.1 249 Intron XP_005270463.1
XM_005270407.1 249 Intron XP_005270464.1
XM_006710312.1 249 Intron XP_006710375.1
XM_011540557.1 249 Intron XP_011538859.1
XM_011540563.1 249 Intron XP_011538865.1
XM_011540566.1 249 Intron XP_011538868.1
XM_017000167.1 249 Intron XP_016855656.1
XM_017000168.1 249 Intron XP_016855657.1
XM_017000169.1 249 Intron XP_016855658.1
XM_017000170.1 249 UTR 5 XP_016855659.1
XM_017000171.1 249 Intron XP_016855660.1
XM_017000172.1 249 Intron XP_016855661.1
XM_017000173.1 249 Intron XP_016855662.1
XM_017000174.1 249 Intron XP_016855663.1
XM_017000175.1 249 Intron XP_016855664.1
XM_017000176.1 249 Intron XP_016855665.1
XM_017000177.1 249 Intron XP_016855666.1
XM_017000178.1 249 Intron XP_016855667.1

View Full Product Details