Product Details
- SNP ID
-
hCV27848870
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55028271 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCACGTGCCCACCTTTCTGTTAG[A/G]GCATTCATCCTGAAGATTTTTATGT
- Phenotype
-
MIM: 600784
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GZMK
PubMed Links
Gene Details
- Gene
- GZMK
- Gene Name
- granzyme K
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002104.2 |
|
Intron |
|
|
NP_002095.1 |
- Gene
- LOC102467081
- Gene Name
- uncharacterized LOC102467081
There are no transcripts associated with this gene.
View Full Product Details