Product Details

SNP ID
rs6763560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:156825917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAACCAGAACTTCCCTTTACAC[C/G]CATGACCTCTACAGGCATCTCTTTT
Phenotype
MIM: 613536
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LEKR1 PubMed Links

Gene Details

Gene
LEKR1
Gene Name
leucine, glutamate and lysine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004316.2 690 Intron NP_001004316.2
NM_001193283.1 690 Intron NP_001180212.1
XM_005247441.3 690 UTR 5 XP_005247498.1
XM_005247442.3 690 UTR 5 XP_005247499.1
XM_006713631.3 690 Intron XP_006713694.1
XM_006713636.2 690 Intron XP_006713699.1
XM_011512803.2 690 Intron XP_011511105.1
XM_011512804.2 690 UTR 5 XP_011511106.1
XM_011512806.2 690 Intron XP_011511108.1
XM_011512807.2 690 Intron XP_011511109.1
XM_011512808.2 690 Intron XP_011511110.1
XM_011512809.1 690 Intron XP_011511111.1
XM_011512811.2 690 Intron XP_011511113.1
XM_011512813.2 690 Intron XP_011511115.1
XM_011512814.2 690 Intron XP_011511116.1
XM_011512815.2 690 Intron XP_011511117.1
XM_011512816.2 690 Intron XP_011511118.1
XM_017006374.1 690 Intron XP_016861863.1
XM_017006375.1 690 Intron XP_016861864.1
Gene
LINC00886
Gene Name
long intergenic non-protein coding RNA 886
There are no transcripts associated with this gene.

View Full Product Details