Product Details

SNP ID
rs7916697
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68232096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAAGGAGCGCTTTTAGCGCAGT[A/G]GCTGTCGGAGAGTGCAGAATGAATC
Phenotype
MIM: 609875
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATOH7 PubMed Links

Gene Details

Gene
ATOH7
Gene Name
atonal bHLH transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145178.3 18 UTR 5 NP_660161.1

View Full Product Details