Product Details
- SNP ID
-
rs17098881
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:61575581 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTGGAAGACAAACAAACAAAAGT[A/C]GTGCCAAACTAAGCATTTGTGATGG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ22447
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs71979984] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ22447
- Gene Name
- uncharacterized LOC400221
There are no transcripts associated with this gene.
- Gene
- LOC101927780
- Gene Name
- uncharacterized LOC101927780
There are no transcripts associated with this gene.
View Full Product Details