Product Details

SNP ID
hCV27852613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75815154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGCCGTTGGGTCCCGCCCCCT[C/T]GAGGGCTAAGCCAATGAGCGCTCCG
Phenotype
MIM: 602493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506113 PubMed Links

Gene Details

Gene
LOC100506113
Gene Name
uncharacterized LOC100506113
There are no transcripts associated with this gene.

Gene
UVRAG
Gene Name
UV radiation resistance associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003369.3 Intron NP_003360.2

View Full Product Details