Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001252119.1 | 107 | Intron | NP_001239048.1 | ||
NM_001252120.1 | 107 | Intron | NP_001239049.1 | ||
NM_001252122.1 | 107 | Intron | NP_001239051.1 | ||
NM_001252124.1 | 107 | Intron | NP_001239053.1 | ||
NM_015148.3 | 107 | Intron | NP_055963.2 | ||
XM_005246991.1 | 107 | Intron | XP_005247048.1 | ||
XM_011510826.1 | 107 | Intron | XP_011509128.1 | ||
XM_011510828.1 | 107 | Intron | XP_011509130.1 | ||
XM_011510829.1 | 107 | UTR 5 | XP_011509131.1 | ||
XM_011510830.1 | 107 | Intron | XP_011509132.1 | ||
XM_011510831.2 | 107 | UTR 5 | XP_011509133.1 | ||
XM_011510832.2 | 107 | UTR 5 | XP_011509134.1 | ||
XM_011510833.1 | 107 | Intron | XP_011509135.1 | ||
XM_011510834.1 | 107 | Intron | XP_011509136.1 | ||
XM_011510835.1 | 107 | Intron | XP_011509137.1 | ||
XM_017003634.1 | 107 | UTR 5 | XP_016859123.1 | ||
XM_017003635.1 | 107 | Intron | XP_016859124.1 | ||
XM_017003636.1 | 107 | UTR 5 | XP_016859125.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282409.1 | 107 | UTR 5 | NP_001269338.1 | ||
NM_001282410.1 | 107 | UTR 5 | NP_001269339.1 | ||
NM_001282411.1 | 107 | UTR 5 | NP_001269340.1 | ||
NM_001282412.1 | 107 | Intron | NP_001269341.1 | ||
NM_001282413.1 | 107 | Intron | NP_001269342.1 | ||
NM_001282414.1 | 107 | Intron | NP_001269343.1 | ||
NM_002712.2 | 107 | UTR 5 | NP_002703.1 | ||
XM_011511388.1 | 107 | Intron | XP_011509690.1 |