Product Details

SNP ID
rs115800751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50924819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTCCTTCTGAGGAAAAGCTGG[A/G]AACCTTCTCATTTTGCCTTATGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL7A PubMed Links

Gene Details

Gene
METTL7A
Gene Name
methyltransferase like 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014033.3 69 UTR 5 NP_054752.3
XM_006719332.1 69 UTR 5 XP_006719395.1

View Full Product Details