Product Details

SNP ID
rs28547082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29193669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGATTCACCACAAGTACATTGG[A/T]GTAACCACTGGGAACAACAGGTTGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details