Product Details

SNP ID
hCV27856847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153904696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGGCTGGGCTGGGCCAGGGCA[G/T]GGCAGCCGATGGAGAGCAGATCTGA
Phenotype
MIM: 300023 MIM: 300538
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
There are no transcripts associated with this gene.

Gene
AVPR2
Gene Name
arginine vasopressin receptor 2
There are no transcripts associated with this gene.

View Full Product Details