Product Details

SNP ID
rs77510571
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:68100588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTTAAATTTAGAAATTAAAAT[C/T]GAATTGTCTAAACTGGCAAAAGTCT
Phenotype
MIM: 615406 MIM: 611514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNG12-AS1 PubMed Links

Gene Details

Gene
GNG12-AS1
Gene Name
GNG12 antisense RNA 1
There are no transcripts associated with this gene.

Gene
WLS
Gene Name
wntless Wnt ligand secretion mediator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002292.3 Intron NP_001002292.3
NM_001193334.1 Intron NP_001180263.1
NM_024911.6 Intron NP_079187.3
XM_011542191.2 Intron XP_011540493.1
XM_011542192.2 Intron XP_011540494.1
XM_017002390.1 Intron XP_016857879.1

View Full Product Details