Product Details
- SNP ID
-
rs8108837
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:32691896 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCCGGAGGTGCTGGGGTCCCTG[C/T]AGGGCCGGGCCACCTGCCGTGGAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUDT19
PubMed Links
Gene Details
- Gene
- NUDT19
- Gene Name
- nudix hydrolase 19
View Full Product Details