Product Details

SNP ID
rs115752470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73830651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATGTTTTCTAGTCTGCCAGTAC[A/T]CTCATCGTTGTGATTTGGGCTTCCT
Phenotype
MIM: 606247
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STAMBP PubMed Links

Gene Details

Gene
STAMBP
Gene Name
STAM binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006463.4 Intron NP_006454.1
NM_201647.2 Intron NP_964010.1
NM_213622.2 Intron NP_998787.1
XM_005264088.3 Intron XP_005264145.1
XM_011532483.2 Intron XP_011530785.1
XM_011532484.2 Intron XP_011530786.1
XM_011532485.2 Intron XP_011530787.1
XM_017003175.1 Intron XP_016858664.1
XM_017003176.1 Intron XP_016858665.1
XM_017003177.1 Intron XP_016858666.1
XM_017003178.1 Intron XP_016858667.1
XM_017003179.1 Intron XP_016858668.1
XM_017003180.1 Intron XP_016858669.1
XM_017003181.1 Intron XP_016858670.1

View Full Product Details