Product Details

SNP ID
rs72878797
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:32364998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGAGGACAAGGTACGTCCTCT[A/G]GACCTAGGCTATCCCCCTACCCTGT
Phenotype
MIM: 607660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BSDC1 PubMed Links

Gene Details

Gene
BSDC1
Gene Name
BSD domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143888.2 2679 UTR 3 NP_001137360.1
NM_001143889.2 2679 UTR 3 NP_001137361.1
NM_001143890.2 2679 UTR 3 NP_001137362.1
NM_001300958.1 2679 UTR 3 NP_001287887.1
NM_018045.7 2679 UTR 3 NP_060515.3
XM_005270986.4 2679 Intron XP_005271043.1
XM_011541677.2 2679 Intron XP_011539979.1
Gene
FAM229A
Gene Name
family with sequence similarity 229 member A
There are no transcripts associated with this gene.

Gene
TSSK3
Gene Name
testis specific serine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052841.3 2679 Intron NP_443073.1

View Full Product Details