Product Details

SNP ID
rs201509501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143320675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTCTTCCTAGATTGTATCCAC[C/T]GCCTGGGACAGGTGGTGAGAAGAAA
Phenotype
MIM: 118425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCN1 PubMed Links

Gene Details

Gene
CLCN1
Gene Name
chloride voltage-gated channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000083.2 258 Missense Mutation CGC,TGC R105C NP_000074.2
XM_011515782.2 258 Intron XP_011514084.1
XM_017011739.1 258 Missense Mutation CCG,CTG P7L XP_016867228.1
XM_017011740.1 258 Missense Mutation CCG,CTG P7L XP_016867229.1

View Full Product Details