Product Details

SNP ID
rs142133355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69035784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGGCGGCAGACCCGGAGCAGCA[C/T]GTGGACTCTCGGGCGCCGCGCAGTA
Phenotype
MIM: 606829
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FXN PubMed Links

Gene Details

Gene
FXN
Gene Name
frataxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000144.4 222 Missense Mutation ACG,ATG T1M NP_000135.2
NM_001161706.1 222 Missense Mutation ACG,ATG T0M NP_001155178.1
NM_181425.2 222 Missense Mutation ACG,ATG T1M NP_852090.1

View Full Product Details