Product Details

SNP ID
rs4871789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:127415816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCACAGAGCTTTCAATGAAAAGTA[A/G]CATAATTGCTCATTTCACCAGGCCC
Phenotype
MIM: 615739
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASC8 PubMed Links

Gene Details

Gene
CASC8
Gene Name
cancer susceptibility candidate 8 (non-protein coding)
There are no transcripts associated with this gene.

Gene
POU5F1B
Gene Name
POU class 5 homeobox 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159542.1 Intron NP_001153014.1

View Full Product Details