Product Details

SNP ID
rs4749206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26940998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGTTTTTACACACTTACCTTAC[C/G]GAAGCCTCATAACAAGCACATCGTT
Phenotype
MIM: 610855
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD26 PubMed Links

Gene Details

Gene
ANKRD26
Gene Name
ankyrin repeat domain 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256053.1 22192 Intron NP_001242982.1
NM_014915.2 22192 Intron NP_055730.2
XM_006717423.2 22192 Intron XP_006717486.1
XM_006717425.3 22192 Intron XP_006717488.1
XM_011519416.2 22192 Intron XP_011517718.1
XM_017015928.1 22192 Intron XP_016871417.1
XM_017015929.1 22192 UTR 3 XP_016871418.1
XM_017015930.1 22192 Intron XP_016871419.1
XM_017015931.1 22192 Intron XP_016871420.1
XM_017015932.1 22192 Intron XP_016871421.1
XM_017015933.1 22192 Intron XP_016871422.1
XM_017015934.1 22192 Intron XP_016871423.1
Gene
LINC00202-1
Gene Name
long intergenic non-protein coding RNA 202-1
There are no transcripts associated with this gene.

View Full Product Details