Product Details

SNP ID
rs4892250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74600513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGAGAGATGGTCTTTATCCCT[C/T]GCAAAGACCCACACCTCAAAGGAGT
Phenotype
MIM: 615894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00909 PubMed Links

Gene Details

Gene
LINC00909
Gene Name
long intergenic non-protein coding RNA 909
There are no transcripts associated with this gene.

Gene
ZNF407
Gene Name
zinc finger protein 407
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146189.1 Intron NP_001139661.1
NM_001146190.1 Intron NP_001139662.1
NM_017757.2 Intron NP_060227.2
XM_005266726.4 Intron XP_005266783.1
XM_006722500.3 Intron XP_006722563.1
XM_011526068.2 Intron XP_011524370.1
XM_011526069.2 Intron XP_011524371.1
XM_011526070.1 Intron XP_011524372.1
XM_017025838.1 Intron XP_016881327.1

View Full Product Details