Product Details

SNP ID
rs4682434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112815222 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAAATAAAATTTCATAGATTTCA[G/T]TTCTCCTGTTTCTCCTACTCACATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD200R1L PubMed Links

Gene Details

Gene
CD200R1L
Gene Name
CD200 receptor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008784.2 2232 Intron NP_001008784.2
NM_001199215.1 2232 Intron NP_001186144.1
XM_011512763.2 2232 UTR 3 XP_011511065.1
XM_017006300.1 2232 UTR 3 XP_016861789.1
Gene
LOC100129297
Gene Name
uncharacterized LOC100129297
There are no transcripts associated with this gene.

View Full Product Details