Product Details

SNP ID
rs4978888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110201422 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGACAGTGAGCTTCTTCTCTCCTT[A/G]TTAATCCAAACAGCATTGACCATCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf152 PubMed Links

Gene Details

Gene
C9orf152
Gene Name
chromosome 9 open reading frame 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012993.2 800 Silent Mutation AAC,AAT N82N NP_001013011.2

View Full Product Details