Product Details
- SNP ID
-
rs4854679
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:133040969 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGATCTGTGTCACATTGTGTAAG[C/T]ATAAGAGCCTTCATATTGAGAGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM108
PubMed Links
Gene Details
- Gene
- TMEM108
- Gene Name
- transmembrane protein 108
View Full Product Details