Product Details

SNP ID
rs4450558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:160107447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCTCAGGAGCCTCATGAGACC[C/T]TTCATCGTGGTGGAAATCCCCACCG
Phenotype
MIM: 147558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGB6 PubMed Links

Gene Details

Gene
ITGB6
Gene Name
integrin subunit beta 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000888.4 Intron NP_000879.2
NM_001282353.1 Intron NP_001269282.1
NM_001282354.1 Intron NP_001269283.1
NM_001282355.1 Intron NP_001269284.1
NM_001282388.1 Intron NP_001269317.1
NM_001282389.1 Intron NP_001269318.1
NM_001282390.1 Intron NP_001269319.1

View Full Product Details