Product Details

SNP ID
rs4589553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2433009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGACAACCACGCCAGTGTGTGG[A/G]CATGTGCCAGCTTCCAGGAGCTGTG
Phenotype
MIM: 600227
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA17P PubMed Links

Gene Details

Gene
ABCA17P
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
There are no transcripts associated with this gene.

Gene
CCNF
Gene Name
cyclin F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323538.1 468 Intron NP_001310467.1
NM_001761.2 468 Missense Mutation ACA,GCA T74A NP_001752.2
XM_017023822.1 468 Missense Mutation ACA,GCA T155A XP_016879311.1
XM_017023823.1 468 Missense Mutation ACA,GCA T155A XP_016879312.1

View Full Product Details