Product Details
- SNP ID
-
rs4851126
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:98356161 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAAGAGTATTTTGGAAAAGTGTTT[C/T]AGAAAAGTTTCAAAGTATGTAGACC
- Phenotype
-
MIM: 600053
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNGA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112708953] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNGA3
- Gene Name
- cyclic nucleotide gated channel alpha 3
View Full Product Details