Product Details

SNP ID
rs4304543
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:37490152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATCAAAGGCCTAATCTTTTCTT[C/T]CCACCCCCACCCAGCCATCCTGTGT
Phenotype
MIM: 611001 MIM: 610562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEAF6 PubMed Links

Gene Details

Gene
MEAF6
Gene Name
MYST/Esa1 associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270875.1 4626 UTR 3 NP_001257804.1
NM_001270876.1 4626 UTR 3 NP_001257805.1
NM_022756.5 4626 UTR 3 NP_073593.2
Gene
MIR6732
Gene Name
microRNA 6732
There are no transcripts associated with this gene.

Gene
ZC3H12A
Gene Name
zinc finger CCCH-type containing 12A
There are no transcripts associated with this gene.

View Full Product Details