Product Details

SNP ID
rs4236645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134781974 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTGACTGGCCTGAGAATCTCCT[G/T]GTCTTAGGTAGATAGAATAAGTTGA
Phenotype
MIM: 114213
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CALD1 PubMed Links

Gene Details

Gene
CALD1
Gene Name
caldesmon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004342.6 Intron NP_004333.1
NM_033138.3 Intron NP_149129.2
NM_033139.3 Intron NP_149130.1
NM_033140.3 Intron NP_149131.1
NM_033157.3 Intron NP_149347.2
XM_017012650.1 Intron XP_016868139.1
XM_017012651.1 Intron XP_016868140.1
XM_017012652.1 Intron XP_016868141.1
XM_017012653.1 Intron XP_016868142.1
XM_017012654.1 Intron XP_016868143.1

View Full Product Details