Product Details
- SNP ID
-
hCV27936614
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:141606155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCAGCTCCTGTATTTTCTCTTTT[C/T]TGGAATGAGTAATTACAATTCTAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPANXA2-OT1
PubMed Links
Gene Details
- Gene
- SPANXA2-OT1
- Gene Name
- SPANXA2 overlapping transcript 1
There are no transcripts associated with this gene.
View Full Product Details