Product Details

SNP ID
rs4833081
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38667197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTCCAGCTGCTCATGAATATA[A/G]GGTCAGGGAGGGCCGGCTAAATGTT
Phenotype
MIM: 609392
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KLF3 PubMed Links

Gene Details

Gene
KLF3
Gene Name
Kruppel like factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016531.5 Intron NP_057615.3
Gene
KLF3-AS1
Gene Name
KLF3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details