Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320238.1 | 3046 | UTR 3 | NP_001307167.1 | ||
NM_001320239.1 | 3046 | UTR 3 | NP_001307168.1 | ||
NM_001320240.1 | 3046 | UTR 3 | NP_001307169.1 | ||
NM_020850.2 | 3046 | UTR 3 | NP_065901.1 | ||
XM_005256070.2 | 3046 | UTR 3 | XP_005256127.1 | ||
XM_005256072.2 | 3046 | Intron | XP_005256129.1 | ||
XM_006721238.3 | 3046 | Intron | XP_006721301.1 | ||
XM_011523245.2 | 3046 | Intron | XP_011521547.1 | ||
XM_011523247.2 | 3046 | Intron | XP_011521549.1 | ||
XM_017023507.1 | 3046 | UTR 3 | XP_016878996.1 | ||
XM_017023508.1 | 3046 | Intron | XP_016878997.1 | ||
XM_017023509.1 | 3046 | UTR 3 | XP_016878998.1 |