Product Details

SNP ID
rs4675634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206738750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAAGCTATTTTTATGTGCTCTCT[C/T]CTGTCACCTTGCTTTTCATTCCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDH1B PubMed Links

Gene Details

Gene
MDH1B
Gene Name
malate dehydrogenase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039845.2 Intron NP_001034934.1
NM_001282940.1 Intron NP_001269869.1
XM_005246315.4 Intron XP_005246372.1
XM_005246316.4 Intron XP_005246373.1
XM_011510628.2 Intron XP_011508930.1
XM_017003373.1 Intron XP_016858862.1
XM_017003374.1 Intron XP_016858863.1

View Full Product Details