Product Details
- SNP ID
-
rs4740382
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:130904626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACATTCCCTCTCCTGCCTCCCCG[A/C]TCCTCGGCCTGAGAGGGGAAACCCA
- Phenotype
-
MIM: 613357
MIM: 609795
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FIBCD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144769166] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FIBCD1
- Gene Name
- fibrinogen C domain containing 1
- Gene
- QRFP
- Gene Name
- pyroglutamylated RFamide peptide
There are no transcripts associated with this gene.
View Full Product Details