Product Details

SNP ID
rs4898990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59506611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGCACAGGGGTTTTTTAAAATC[G/T]CCAGGTGATTCTAATGTTTGAGAAC
Phenotype
MIM: 611176
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC175 PubMed Links

Gene Details

Gene
CCDC175
Gene Name
coiled-coil domain containing 175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164399.1 Intron NP_001157871.1
XM_011537120.2 Intron XP_011535422.1
XM_011537122.2 Intron XP_011535424.1
XM_011537124.2 Intron XP_011535426.1
XM_011537125.2 Intron XP_011535427.1
XM_011537130.2 Intron XP_011535432.1
XM_017021621.1 Intron XP_016877110.1
Gene
JKAMP
Gene Name
JNK1/MAPK8-associated membrane protein
There are no transcripts associated with this gene.

View Full Product Details