Product Details

SNP ID
rs4389136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53713946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTATGAGTTTGACTATTAGACGT[A/G]CATTTTTGTTTTTGCTATAAGCATC
Phenotype
MIM: 610966
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTO PubMed Links

Gene Details

Gene
FTO
Gene Name
fat mass and obesity associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080432.2 Intron NP_001073901.1
XM_011523313.2 Intron XP_011521615.1
XM_011523314.2 Intron XP_011521616.1
XM_011523315.2 Intron XP_011521617.1
XM_011523316.2 Intron XP_011521618.1
XM_017023654.1 Intron XP_016879143.1
XM_017023655.1 Intron XP_016879144.1
XM_017023656.1 Intron XP_016879145.1
XM_017023657.1 Intron XP_016879146.1
XM_017023658.1 Intron XP_016879147.1

View Full Product Details