Product Details
- SNP ID
-
rs4766397
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:1618673 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTCTTTCTTATCATCTGCACAGA[A/G]TAAGACTTCCAGTGGACTTTTGCTT
- Phenotype
-
MIM: 606361
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WNT5B
PubMed Links
Gene Details
- Gene
- WNT5B
- Gene Name
- Wnt family member 5B
View Full Product Details