Product Details

SNP ID
rs4970714
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:92475800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCTATTTGTGTCCGAAGCCTAG[A/G]GAGTCACTTGGTTCTCACTCTCGGC
Phenotype
MIM: 600871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFI1 PubMed Links

Gene Details

Gene
GFI1
Gene Name
growth factor independent 1 transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127215.1 1866 UTR 3 NP_001120687.1
NM_001127216.1 1866 UTR 3 NP_001120688.1
NM_005263.3 1866 UTR 3 NP_005254.2
XM_005270749.3 1866 UTR 3 XP_005270806.1
XM_011541245.2 1866 UTR 3 XP_011539547.1
XM_011541246.2 1866 UTR 3 XP_011539548.1

View Full Product Details