Product Details

SNP ID
rs4975638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1395584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACTGCAGGCCAGGGGTCTACC[A/G]GGGGCCTGGAGCAGCCACAGCTGAT
Phenotype
MIM: 126455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A3 PubMed Links

Gene Details

Gene
SLC6A3
Gene Name
solute carrier family 6 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044.4 Intron NP_001035.1

View Full Product Details