Product Details
- SNP ID
-
hCV27998070
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141973473 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTGGGTAAGCTGGATAGCACTCA[G/T]GAACAGCAGTCCATGCAGGAAAAGC
- Phenotype
-
MIM: 607751
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R38
PubMed Links
Gene Details
- Gene
- TAS2R38
- Gene Name
- taste 2 receptor member 38
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_176817.4 |
301 |
Missense Mutation |
ATG,CTG |
M73L |
NP_789787.4 |
View Full Product Details