Product Details
- SNP ID
-
rs4783971
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57100822 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCTTGCAGCTGACAGGGTGGGG[C/G]TGGGCTCTCTGAGCTCCCACACTTA
- Phenotype
-
MIM: 604206
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE2
PubMed Links
Gene Details
- Gene
- CPNE2
- Gene Name
- copine 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152727.5 |
|
Intron |
|
|
NP_689940.3 |
View Full Product Details