Product Details

SNP ID
rs4547041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84182687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCATCTCTACCACATTCATT[C/T]TGGACAAAACAAGTTGCAAGTCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf99 PubMed Links

Gene Details

Gene
C10orf99
Gene Name
chromosome 10 open reading frame 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207373.2 Intron NP_997256.1

View Full Product Details