Product Details

SNP ID
rs4774022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:106810005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTCCCAATCTGAATCCCAAAG[C/G]CTGTTCCAATCAGAGATTCCCACGG
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C14orf99 PubMed Links

Gene Details

Gene
C14orf99
Gene Name
uncharacterized LOC317730
There are no transcripts associated with this gene.

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

View Full Product Details