Product Details

SNP ID
rs4691944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155832022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGTTTTATATGCTATTTAATGT[C/T]AAACTGACACAGAATTGTTAATATT
Phenotype
MIM: 616693
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASIC5 PubMed Links

Gene Details

Gene
ASIC5
Gene Name
acid sensing ion channel subunit family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017419.2 Intron NP_059115.1
XM_017008291.1 Intron XP_016863780.1

View Full Product Details